Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha-2-macroglobulin Mouse anti-Human, Clone: 4B11F7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Alpha-2-macroglobulin |
---|---|
Clone | 4B11F7 |
Concentration | 1.8 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6562
![]() ![]() |
Proteintech Group Inc
661261IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Alpha 2-Macroglobulin (a2M), is a approximately 720-kDa homotetrameric glycoprotein composed of four identical approximately 180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregcy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer′s disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.Specifications
Alpha-2-macroglobulin | |
1.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2M, Alpha 2 M, alpha 2 macroglobulin, CPAMD5, FWP007, S863 7, alpha-2M | |
A2M | |
IgG2a | |
Protein A | |
Antibody |
4B11F7 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P01023 | |
2 | |
Alpha-2-macroglobulin Fusion Protein Ag19245 | |
Primary | |
-20°C | |
A2M |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title