Learn More
alpha-2-Macroglobulin Polyclonal Antibody, HRP, Rockland™

Goat Polyclonal Antibody
$178.35 - $638.37
Specifications
| Antigen | alpha-2-Macroglobulin |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0352
![]()
|
Rockland Immunochemicals
2001032070100 |
100 μg |
Each for $638.37
|
|
|||||
50-199-0353
![]()
|
Rockland Immunochemicals
200103207S |
25 μL |
Each for $178.35
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Alpha 2-Macroglobulin is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Goat Serum as well as purified and partially purified a2-Macroglobulin [Human Plasma]. Cross reactivity against a2-Macroglobulin from other sources is unknown.
Alpha 2-Macroglobulin (a2M), is a approximately 720-kDa homotetrameric glycoprotein composed of four identical approximately 180 kDa subunits. aAlpha 2M shares with other alpha-macroglobulins, like the complement components C3 and C4 and the pregnancy zone protein PZP, an extraordinary binding capacity for a variety of ligands. This allows the alpha-macroglobulins to serve as humoral defense barriers against foreign peptides in the plasma. Alpha 2M interacts and captures virtually any proteinase, often referred to as a panprotease inhibitor. In the brain of Alzheimer's disease (AD) patients, Alpha 2M also has been localized to diffuse amyloid plaques, supporting an important role for alpha 2M in AD etiopathology.Specifications
| alpha-2-Macroglobulin | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| HRP | |
| Goat | |
| Human | |
| P01023 | |
| 2 | |
| a2-Macroglobulin [Human Plasma] | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| A2M |
| 1 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
| A2m; A2m1; A2maa; A2MAC1; A2MD; A2mp; Alpha 2 M; Alpha-2-M; Alpha-2-macroglobulin; Alpha-2-macroglobulin-P; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5; CPAMD5; DKFZp779B086; FWP007; Mam; S863 7; S863-7 | |
| A2M | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.