Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-2A Adrenergic R/ADRA2A Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1002819
Description
alpha-2A Adrenergic R/ADRA2A Polyclonal specifically detects alpha-2A Adrenergic R/ADRA2A in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
alpha-2A Adrenergic R/ADRA2A | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ADRA2, ADRA2Ralpha-2-adrenergic receptor, platelet type, ADRARZNF32, adrenergic, alpha-2A-, receptor, Alpha-2 adrenergic receptor subtype C10, alpha-2A adrenergic receptor, Alpha-2A adrenoceptor, Alpha-2A adrenoreceptor, Alpha-2AAR, ALPHA2AAR, alpha-2AAR subtype C10 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Mouse. Expected from sequence similarity: Human, Rat and Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000 | |
NP_000672.2 | |
ADRA2A | |
Peptide with sequence C-TERRPNGLGPERS corresponding to internal region according to NP_000672.2. | |
0.1 mg | |
GPCR | |
150 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction