Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-2a Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518475
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518475 Supplier Invitrogen™ Supplier No. PA518475
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-2a Adrenergic Receptor
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Adra2a
Gene Accession No. P08913
Gene Alias ADRA2; Adra-2; ADRA2A; Adra-2a; ADRA2R; ADRAR; adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; adrenoceptor alpha 2A; alpha(2A)AR; alpha-2 adrenergic receptor subtype C10; alpha2A; alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; alpha-2A adrenoreceptor; alpha2A-adrenergic receptor; ALPHA2AAR; alpha-2AAR; alpha2A-AR; alpha-2AAR subtype C10; alpha-2-adrenergic receptor, platelet type; alpha2-C10; alpha-2D adrenergic receptor; AW122659; CA2-47; OTTHUMP00000058849; RATRG20; RG20; subtype alpha2-C10; ZNF32
Gene Symbols Adra2a
Host Species Goat
Immunogen Synthetic peptide sequence (TERRPNGLGPERS) corresponding to the internal amino acids of ADRA2A (aa 261-274).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.