Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha-2a Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114152 Supplier Invitrogen™ Supplier No. PA5114152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRPERGPRGK GKARASQVKP GDSLPRRGPG ATGIGTPAAG PGEERVGAAK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 85%; Guinea Pig: 79%; Human: 100%; Mouse: 79%; Pig: 85%; Rat: 79%.

Alpha-2-adrenergic receptors (ADRA2A) are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. ADRA2A mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G protein. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha-2a Adrenergic Receptor
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Adra2a
Gene Accession No. P08913
Gene Alias ADRA2; Adra-2; ADRA2A; Adra-2a; ADRA2R; ADRAR; adrenergic receptor, alpha 2a; adrenergic, alpha-2A-, receptor; adrenoceptor alpha 2A; alpha(2A)AR; alpha-2 adrenergic receptor subtype C10; alpha2A; alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; alpha-2A adrenoreceptor; alpha2A-adrenergic receptor; ALPHA2AAR; alpha-2AAR; alpha2A-AR; alpha-2AAR subtype C10; alpha-2-adrenergic receptor, platelet type; alpha2-C10; alpha-2D adrenergic receptor; AW122659; CA2-47; OTTHUMP00000058849; RATRG20; RG20; subtype alpha2-C10; ZNF32
Gene Symbols Adra2a
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Human ADRA2A (aa 324-373).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.