Learn More
alpha-2B Adrenergic R/ADRA2B Antibody (491613) [Allophycocyanin], Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals FAB10324A
Catalog No. NB401874
Only null left
Add to cart
Description
Description
alpha-2B Adrenergic R/ADRA2B Monoclonal antibody specifically detects alpha-2B Adrenergic R/ADRA2B in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
alpha-2B Adrenergic R/ADRA2B
Monoclonal
APC
PBS
Mouse
Protein A or G purified from ascites
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
491613
Flow Cytometry, CyTOF-ready
ADRA2B adrenergic, alpha-2B-, receptor, ADRA2L1adrenergic receptor alpha 2B, ADRA2RL1, ADRARL1, adrenergic, alpha-2B-, receptor, Alpha-2 adrenergic receptor subtype C2, alpha-2-adrenergic receptor-like 1, alpha-2B adrenergic receptor, Alpha-2B adrenoceptor, Alpha-2B adrenoreceptor, alpha-2B-adrenergic receptor, Alpha-2BAR, ALPHA2BAR, G-protein coupled receptor
NS0 mouse myeloma cell line transfected with human alpha-2B Adrenergic R/ADRA2B, Accession # P18089
0.1 mL
Adaptive Immunity, GPCR, Immunology, Innate Immunity
151
Store at 4°C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.