Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2b Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549610
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549610 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549610 Supplier Thermo Scientific Supplier No. PA549610
Add to Cart
Add to Cart

alpha-2b Adrenergic Receptor Polyclonal Antibody

alpha-2b Adrenergic Receptor Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-2b Adrenergic Receptor
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADRA2B
Gene Accession No. P18089
Gene Alias [a]2B; ADA2B; Adra2b; Adra-2b; ADRA2B adrenergic, alpha-2B-, receptor; ADRA2L1; ADRA2RL1; ADRARL1; Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2b; adrenergic, alpha-2B-, receptor; Adrenergic, alpha2B-, receptor class III; adrenoceptor alpha 2B; alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; alpha2B; alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; alpha-2B adrenoreceptor; ALPHA2BAR; alpha-2BAR; alpha2-C2; FAME2; G-protein coupled receptor; subtype alpha2-C2
Gene Symbols ADRA2B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 343-369 amino acids from the Central region of human ADRA2B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.