Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-2b Adrenergic Receptor Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA564258

Catalog No. PIPA564258


Only null left
Add to Cart

Description

Description

Immunogen sequence: IAKRSNRRGP RAKGGPGQGE SKQPRPDHGG ALASAKLPAL ASVASAREVN Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 55%.

Alpha-2-adrenergic receptors are members of the G protein-coupled receptor superfamily. They include 3 highly homologous subtypes: alpha2A, alpha2B, and alpha2C. These receptors have a critical role in regulating neurotransmitter release from sympathetic nerves and from adrenergic neurons in the central nervous system. Alpha 2B adrenergic receptor subtype was observed to associate with eIF-2B, a guanine nucleotide exchange protein that functions in regulation of translation. A polymorphic variant of the alpha2B subtype, which lacks 3 glutamic acids from a glutamic acid repeat element, was identified to have decreased G protein-coupled receptor kinase-mediated phosphorylation and desensitization; this polymorphic form is also associated with reduced basal metabolic rate in obese subjects and may therefore contribute to the pathogenesis of obesity. Alpha 2B adrenergic receptor gene contains no introns in either its coding or untranslated sequences.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

alpha-2b Adrenergic Receptor
Polyclonal
Unconjugated
ADRA2B
[a]2B; ADA2B; Adra2b; Adra-2b; ADRA2B adrenergic, alpha-2B-, receptor; ADRA2L1; ADRA2RL1; ADRARL1; Adrenergic alpha2B- receptor class III; adrenergic receptor, alpha 2b; adrenergic, alpha-2B-, receptor; Adrenergic, alpha2B-, receptor class III; adrenoceptor alpha 2B; alpha-2 adrenergic receptor subtype C2; alpha-2-adrenergic receptor-like 1; alpha2B; alpha-2B adrenergic receptor; Alpha-2B adrenoceptor; alpha-2B adrenoreceptor; ALPHA2BAR; alpha-2BAR; alpha2-C2; FAME2; G-protein coupled receptor; subtype alpha2-C2
Rabbit
Antigen Affinity Chromatography
RUO
151
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P18089
ADRA2B
Recombinant protein corresponding to Human alpha-2b Adrenergic Receptor. Recombinant protein control fragment (Product #RP-103896).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.