Learn More
Alpha 2b Adrenergic Receptor Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Alpha 2b Adrenergic Receptor |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | ADRA2B,ADRA2L1,ADRA2RL1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 14 amino acids from the internal region of human alpha-2B adrenergic receptor. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.