Learn More
Invitrogen™ alpha-2c Adrenergic Receptor Monoclonal Antibody (N330A/80)

Description
1 μg/mL of MA5-27724 was sufficient for detection of Alpha2C Adrenergic Receptor in 20 μg of COS cells transiently transfected with HA-tagged Alpha- lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects 50kDa or larger (possibly due to dimerization). Does not cross-react with other adrenergic receptors. This antibody was formerly sold as clone S330A-80.
Specifications
Specifications
| Antigen | alpha-2c Adrenergic Receptor |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N330A/80 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene | ADRA2C |
| Gene Accession No. | P18825, Q01337 |
| Gene Alias | [a]2C; Adra 2c; ADRA2C; Adra-2c; ADRA2L2; ADRA2RL2; ADRARL2; Adrenergic alpha2C- receptor class I; adrenergic receptor, alpha 2c; adrenergic, alpha-2C-, receptor; Adrenergic, alpha2C-, receptor class I; adrenoceptor alpha 2C; Alpha 2CAR; alpha-2 adrenergic receptor subtype C4; alpha2 AR C4; alpha2 C4; alpha2-AR-C4; alpha2C; Alpha-2C adrenergic receptor; alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha2-C4; alpha-2C-adrenergic receptor; ALPHA2CAR; alpha-2CAR; Alpha-2C-AR; subtype alpha2-C4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.