Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-2c Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533330
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533330 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533330 Supplier Thermo Scientific Supplier No. PA533330
Add to Cart
Add to Cart

alpha-2c Adrenergic Receptor Polyclonal Antibody

alpha-2c Adrenergic Receptor Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-2c Adrenergic Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADRA2C
Gene Accession No. P18825
Gene Alias [a]2C; Adra 2c; ADRA2C; Adra-2c; ADRA2L2; ADRA2RL2; ADRARL2; Adrenergic alpha2C- receptor class I; adrenergic receptor, alpha 2c; adrenergic, alpha-2C-, receptor; Adrenergic, alpha2C-, receptor class I; adrenoceptor alpha 2C; Alpha 2CAR; alpha-2 adrenergic receptor subtype C4; alpha2 AR C4; alpha2 C4; alpha2-AR-C4; alpha2C; Alpha-2C adrenergic receptor; alpha-2C adrenoceptor; Alpha-2C adrenoreceptor; alpha2-C4; alpha-2C-adrenergic receptor; ALPHA2CAR; alpha-2CAR; Alpha-2C-AR; subtype alpha2-C4
Gene Symbols ADRA2C
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminal extracellular domain of human ADRA2C.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.