Learn More
Invitrogen™ Alpha A Crystallin Monoclonal Antibody (1H3.B8), APC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545172
Description
0.5 μg/mL was sufficient for detection of 100 ng purified alphaA crystalline by colorimetric immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary. Detects approximately 20kDa. Does not cross-react with alpha B-crystallin, beta L-crystallin, beta H- crystallin, gamma -crystallin, HSP25, HSP27 or HSP47 proteins.
Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.
Specifications
Alpha A Crystallin | |
Monoclonal | |
1 mg/mL | |
2.48 mM MES, 95.64 mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
P02470, P02489, P24622, P24623 | |
CRYAA, CRYAA2 | |
Native Alpha Crystallin. | |
200 μg | |
Primary | |
Human, Mouse, Rat, Bovine | |
Antibody | |
IgG1 |
ELISA, Western Blot, Immunocytochemistry | |
1H3.B8 | |
APC | |
CRYAA | |
[a]A-crystallin; Acry 1; Acry-1; alpha A crystallin; alpha crystallin A chain; alpha-A-crystallin; alpha-crystallin A chain; Alpha-crystallin A(1-151); Alpha-crystallin A(1-156); Alpha-crystallin A(1-157); Alpha-crystallin A(1-162); Alpha-crystallin A(1-163); Alpha-crystallin A(1-165); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); CRYA 1; Crya1; Crya-1; CRYAA; CRYAA2; Crystallin Alpha 1; crystallin alpha A; crystallin, alpha 1; crystallin, alpha A; Crystallin, alpha polypeptide A; crystallin, alpha-1; CTRCT9; DAcry-1; Heat shock protein beta-4; HSPB 4; HSPB4; human alphaA-crystallin (CRYA1); lens opacity 18; LOC102724652; lop18; small heat shock protein; wu:fc06h04; zgc:92036 | |
Mouse | |
Protein G | |
RUO | |
102724652, 12954, 1409, 24273, 281718 | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.