Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha A Crystallin Recombinant Rabbit Monoclonal Antibody (1T1F5)
GREENER_CHOICE

Catalog No. PIMA535646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535646 Supplier Invitrogen™ Supplier No. MA535646
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: QHPWFKRTLG PFYPSRLFDQ FFGEGLFEYD LLPFLSSTIS PYYRQSLFRT VLDSGISEVR SDRDKFVIFL DVKHFSPEDL TVKVQDDFVE.

Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha A Crystallin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1T1F5
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRYAA
Gene Accession No. P24622, P24623
Gene Alias [a]A-crystallin; Acry 1; Acry-1; alpha A crystallin; alpha crystallin A chain; alpha-A-crystallin; alpha-crystallin A chain; Alpha-crystallin A(1-151); Alpha-crystallin A(1-156); Alpha-crystallin A(1-157); Alpha-crystallin A(1-162); Alpha-crystallin A(1-163); Alpha-crystallin A(1-165); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); CRYA 1; Crya1; Crya-1; CRYAA; CRYAA2; Crystallin Alpha 1; crystallin alpha A; crystallin, alpha 1; crystallin, alpha A; Crystallin, alpha polypeptide A; crystallin, alpha-1; CTRCT9; DAcry-1; Heat shock protein beta-4; HSPB 4; HSPB4; human alphaA-crystallin (CRYA1); lens opacity 18; LOC102724652; lop18; small heat shock protein; wu:fc06h04; zgc:92036
Gene Symbols CRYAA
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12954, 24273
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.