Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Alpha A Crystallin Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1009

Catalog No. PA1009


Only null left
Edge
Add to Cart

Description

Description

PA1-009 detects alpha-A crystallin from bovine and rat samples. This antibody does not detect the unphosphorylated form of the protein. PA1-009 has been successfully used in Western blot and immunohistochemistry procedures. By Western blot, this antibody detects an ~20 kDa protein representing alpha-A crystallin from bovine lens samples. Immunohistochemical staining of alpha-A crystallin in rat thymus samples results in staining of epithelial reticular cells. The PA1-009 immunogen is purified alpha-A Crystallin from bovine lens. Reconstitute in 100 μL PBS to create a stock of 1 mg/mL.

Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alpha A Crystallin
Polyclonal
Unconjugated
CRYAA
[a]A-crystallin; Acry 1; Acry-1; alpha A crystallin; alpha crystallin A chain; alpha-A-crystallin; alpha-crystallin A chain; Alpha-crystallin A(1-151); Alpha-crystallin A(1-156); Alpha-crystallin A(1-157); Alpha-crystallin A(1-162); Alpha-crystallin A(1-163); Alpha-crystallin A(1-165); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); CRYA 1; Crya1; Crya-1; CRYAA; CRYAA2; Crystallin Alpha 1; crystallin alpha A; crystallin, alpha 1; crystallin, alpha A; Crystallin, alpha polypeptide A; crystallin, alpha-1; CTRCT9; DAcry-1; Heat shock protein beta-4; HSPB 4; HSPB4; human alphaA-crystallin (CRYA1); lens opacity 18; LOC102724652; lop18; small heat shock protein; wu:fc06h04; zgc:92036
Rabbit
Antigen affinity chromatography
RUO
24273, 281718
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 30mg/mL BSA and 0.05% sodium azide
P02470, P24623
CRYAA
Purified alpha-A Crystallin from bovine lens.
100 μg
Primary
Rat, Bovine
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.