Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Actin-1 Rabbit anti-Bovine, Fish, Human, Mouse, Porcine, Rabbit, Rat, Polyclonal, GenScript™

Catalog No. 50273025
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-025 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-025 Supplier Genscript Corporation Supplier No. A0088540

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL, if the antibody is in lyophilized form. The two major cytoskeletal proteins implicated in cell motility are actin and myosin. Actin and myosin are involved in a myriad of cellular processes including locomotion, secretion, cytoplasmic streaming, phagocytosis, and cytokinesis.GenScript Rabbit Anti-Alpha-Actin-1 Polyclonal Antibody is developed in rabbit hosts using a synthetic peptide conjugated to KLH.

alpha-Actin-1 Polyclonal antibody specifically detects alpha-Actin-1 in Bovine, Fish, Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for Western Blot

Specifications

Antigen alpha-Actin-1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene ACTA1
Gene Accession No. P68133, P68134, P68135, P68136, P68137, P68138
Gene Alias AA959943; ACTA; Acta1; Acta-2; actin alpha 1; actin alpha 1, skeletal muscle; actin, alpha 1, skeletal muscle; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Acts; Actsk-1; alpha actin 1; alpha-actin-1; ASMA; beta actin; CFTD; CFTD1; CFTDM; cytoplasmic beta-actin; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; MPFD; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; skeletal alpha actin; unnamed protein product
Host Species Rabbit
Immunogen Synthetic peptide GQVITIGNERFR conjugated to KLH
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009506, 100154254, 11459, 281592, 29437, 58
Target Species Bovine, Fish, Human, Mouse, Pig, Rabbit, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.