Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ alpha-Actin, Cardiac Muscle, Mouse, Unlabeled, Clone: 22D3,

Catalog No. MABT823MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT823MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT823MI Supplier MilliporeSigma™ Supplier No. MABT823
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Actin, alpha cardiac muscle 1 (UniProt P68032; also known as Alpha-cardiac actin) is encoded by the ACTC1 (also known as ACTC, ASD5, CMD1R, CMH11, LVNC4) gene (Gene ID 70) in human. Actins are globular multi-functional proteins that serve as the basic building blocks of cytoskeletal microfilaments and are among the most conserved eukaryotic proteins. Six actin types exist, skeletal muscle alpha-actin is encoded by the ACTA1 gene, smooth muscle alpha-actin by the ACTA2 gene, cytoplasmic beta-actin by the ACTB gene, cardiac muscle alpha-actin by the ACTC1 gene, cytoplasmic gamma-actin by the ACTG1 gene, and smooth muscle gamma-actin by the ACTG2 (a.k.a. ACTA3) gene. Although actins show >90% overall sequence homology, isoforms do show spatial, temporal, and tissue-specific expresson patterns and only 50-60% homology is found in their 18 N-terminal residues. Cytoplasmic - and -actins, are thought to be present in all cells, while the other four actin isoforms are typically found in specific adult muscle tissue types. Actins exist in a variety of structural states, depending on the specific ionic conditions or the interaction with ligand proteins. The oligomeric and polymeric forms that actin molecules assume are dependent on the distinct conformations they adopt.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Actin, Cardiac Muscle
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 22D3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene ACTC1, ACTC, ASD5, CMD1R, CMH11, LVNC4
Gene Accession No. P68032
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human mature alpha-cardiac actin N-terminal end sequence with an acetylated N-terminus.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_005150.1
Test Specificity Clone 22D3 specifically detected alpha-cardiac actin in rat myocardium homogenates with no cross-reactivity toward - or -smooth muscle actin in rat aorta and chicken gizzard homogenates, cytoplasmic - or -actin in human platelet lysate, nor -skeletal actin in rat skeletal muscle homogenate (Driesen, R.B., et al. (2009). J. Cell. Mol. Med. 13(5):896-908).
Target Species Goat
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.