Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Actinin 1/2/3/4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114820 Supplier Thermo Scientific Supplier No. PA5114820
Add to Cart
Add to Cart

alpha Actinin 1/2/3/4 Polyclonal Antibody

alpha Actinin 1/2/3/4 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Actinin 1/2/3/4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ACTN1
Gene Accession No. O43707, O88990, P12814, P35609, P57780, Q08043, Q7TPR4, Q9JI91, Q9QXQ0, Q9Z1P2
Gene Alias 1110008F24Rik; 3110023F10Rik; actinin 1 smooth muscle; actinin alpha 1; actinin alpha 2; actinin alpha 3; actinin alpha 3 (gene/pseudogene); actinin alpha 4; actinin, alpha 1; actinin, alpha 2; actinin, alpha 3; ACTININ-4; ACTN1; Actn1a; ACTN2; Actn3; Actn4; alpha actinin 1a; alpha actinin 4; alpha-actinin cytoskeletal isoform; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-1; alpha-actinin-2; Alpha-actinin-3; alpha-actinin-4; BDPLT15; C77391; CMD1AA; CMH23; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; non-muscle alpha-actinin 1; non-muscle alpha-actinin 4; Non-muscle alpha-actinin-1
Gene Symbols ACTN1, Actn2, Actn3, Actn4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACTN1/2/3/4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109711, 11472, 11474, 171009, 291245, 60595, 63836, 81, 81634, 87, 88, 89
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.