Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Actinin 1 Antibody (1D10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259465F
This item is not returnable.
View return policy
Description
alpha-Actinin 1 Monoclonal antibody specifically detects alpha-Actinin 1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
alpha-Actinin 1 | |
Monoclonal | |
FITC | |
actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
Recombinant human alpha-Actinin 1 (1-249aa) purified from E. coli | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers | |
87 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry, ELISA, Immunofluorescence | |
1D10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction