Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-Actinin 1 Antibody (OTI7A4), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270078V
Description
alpha-Actinin 1 Monoclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
alpha-Actinin 1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACTN1 | |
Recombinant protein fragment corresponding to amino acids 650-893 of human alpha-Actinin 1 (NP_001093) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
OTI7A4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cancer, Cytoskeleton Markers | |
87.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction