Learn More
Invitrogen™ alpha Actinin 2/3 Polyclonal Antibody

Description
The antibody detects endogenous levels of total alpha Actinin 2 + alpha Actinin 3 protein.
Specifications
Specifications
| Antigen | alpha Actinin 2/3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ACTN2 |
| Gene Accession No. | O88990, P35609, Q08043, Q9JI91 |
| Gene Alias | 1110008F24Rik; actinin alpha 2; actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 2; actinin, alpha 3; Actinin-alpha3; ACTN2; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin skeletal muscle isoform 3; Alpha-actinin-2; alpha-Actinin3; alpha-actinin-3; alphaActinin-3; CMD1AA; CMH23; F-actin cross-linking protein |
| Gene Symbols | ACTN2, ACTN3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.