Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha Actinin 2 Antibody (ACTN2/3292), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB010777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010777 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010777 Supplier Novus Biologicals Supplier No. NBP279838AF700
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Alpha Actinin 2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ACTN2/3292
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein
Gene Symbols ACTN2
Host Species Mouse
Immunogen A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) (Uniprot: P35609)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.