Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 2 Antibody (ACTN2/3293), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279839AF405
Description
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Alpha Actinin 2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ACTN2 | |
A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) (Uniprot: P35609) | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
ACTN2/3293 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction