Learn More
Invitrogen™ alpha Actinin 2 Monoclonal Antibody (EA-53)

Description
Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: rat cardiac muscle tissue, rat skeletal muscle tissue, mouse cardiac muscle tissue, mouse skeletal muscle tissue, human placenta tissue. IHC: human mammary cancer tissue, mouse lung tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | alpha Actinin 2 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | EA-53 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide |
| Gene | Actn2 |
| Gene Accession No. | P35609, Q9JI91 |
| Gene Alias | 1110008F24Rik; actinin alpha 2; actinin, alpha 2; ACTN2; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin-2; CMD1AA; CMH23; F-actin cross-linking protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.