Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Actinin 2 Polyclonal Antibody

Supplier: Thermo Scientific PA5114821
Description
Specifications
| alpha Actinin 2 | |
| Polyclonal | |
| Unconjugated | |
| Actn2 | |
| 1110008F24Rik; actinin alpha 2; actinin, alpha 2; ACTN2; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin-2; CMD1AA; CMH23; F-actin cross-linking protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11472, 291245, 88 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P35609, Q9JI91 | |
| Actn2 | |
| A synthesized peptide derived from human ACTN2(Accession P35609), corresponding to amino acid residues K38-D88. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction