Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 3 Rabbit anti-Human, Mouse, Rat, Clone: 4H7E6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Alpha Actinin 3 |
---|---|
Clone | 4H7E6 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Alpha Actinin 3 Monoclonal antibody specifically detects Alpha Actinin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Alpha Actinin 3 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
89 | |
IgG | |
Affinity purified |
4H7E6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
actinin, alpha 3, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 3, alpha-actinin-3, F-actin cross-linking protein, MGC117002, MGC117005 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 700-800 of human Alpha Actinin 3 (Q08043). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title