Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Actinin 3 Recombinant Rabbit Monoclonal Antibody (4H7E6)
GREENER_CHOICE

Catalog No. PIMA542718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542718 Supplier Invitrogen™ Supplier No. MA542718
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse skeletal muscle, Rat skeletal muscle. Immunogen sequence: DRLEGDHQLL QESLVFDNKH TVYSMEHIRV GWEQLLTSIA RTINEVENQV LTRDAKGLSQ EQLNEFRASF NHFDRKQNGM MEPDDFRACL ISMGYDLGEV E.

Alpha-actinin-3, also known as alpha-actinin skeletal muscle isoform 3 or F-actin cross-linking protein, is a protein that in humans is encoded by the ACTN3 gene. This gene encodes a member of the alpha-actin binding protein gene family. The encoded protein is primarily expressed in skeletal muscle and functions as a structural component of sarcomeric Z line. This protein is involved in crosslinking actin containing thin filaments. An allelic polymorphism in this gene results in both coding and non-coding variants; the reference genome represents the coding allele.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Actinin 3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4H7E6
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Actn3
Gene Accession No. O88990, Q08043
Gene Alias actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 3; Actinin-alpha3; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; alpha-Actinin3; Alpha-actinin-3; alphaActinin-3; F-actin cross-linking protein
Gene Symbols Actn3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 700-800 of human ACTN3 (Q08043).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11474, 171009, 89
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.