Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 4 Antibody (93), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308415AF350
Description
Alpha Actinin 4 Monoclonal antibody specifically detects Alpha Actinin 4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Alpha Actinin 4 | |
Monoclonal | |
Alexa Fluor 350 | |
actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
Recombinant full-length human Alpha Actinin 4 protein (Uniprot: O43707) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
93 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
81 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction