Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323332
Description
Alpha Actinin 4 Polyclonal antibody specifically detects Alpha Actinin 4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Alpha Actinin 4 | |
Polyclonal | |
Western Blot 1:500 | |
actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
Alpha Actinin 4 synthetic peptide (coupled to carrier protein) corresponds to amino acids in the N-terminus of human alpha actinin 4. This sequence is well conserved in rat and mouse alpha actinin 4, but is not conserved in other alpha actinins. [UniProt# O43707] | |
0.1 mL | |
Phospho Specific | |
81 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction