Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Actinin 4 Monoclonal Antibody (7H6)
GREENER_CHOICE

Catalog No. MA1800
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1800 Supplier Invitrogen™ Supplier No. MA1800
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-800 detects the alpha Actinin 4 in human and mouse cells. MA1-800 has been successfully used in Western blot, and immunofluorescence procedures. MA1-800 detects alpha Actinin 4 which has a predicted molecular weight of approximately 105 kDa. Under reducing conditions, MA1-800 detects a monomer of alpha Actinin 4 at approximately 52.5 kDa. The MA1-800 immunogen is a synthetic peptide corresponding to residues A(884) P Y Q G P D A V P G A L D(897) of human alpha Actin 4.

Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Actinin 4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Actn4
Gene Accession No. O43707, P57780
Gene Alias actinin alpha 4; ACTININ-4; actinin-Alpha-4; Actn4; alpha actinin 4; alpha-actinin-4; C77391; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; non-muscle alpha-actinin 4
Gene Symbols Actn4
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues A(884) P Y Q G P D A V P G A L D(897) of human alpha Actinin 4.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60595, 81
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.