Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 4 Rabbit anti-Human, Mouse, Rat, Clone: 5Y3R6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316176100UL
This item is not returnable.
View return policy
Description
Alpha Actinin 4 Monoclonal antibody specifically detects Alpha Actinin 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
Alpha Actinin 4 | |
Monoclonal | |
Unconjugated | |
actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Alpha Actinin 4 (O43707). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
5Y3R6 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
81 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction