Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha Actinin 4 Rabbit anti-Human, Mouse, Rat, Clone: S06-3D7, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | Alpha Actinin 4 |
---|---|
Clone | S06-3D7 |
Dilution | Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence, Immunofluorescence 20 ug/ml |
Applications | Western Blot, Immunofluorescence, Immunofluorescence |
Classification | Monoclonal |
Description
Alpha Actinin 4 Monoclonal antibody specifically detects Alpha Actinin 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunofluorescenceSpecifications
Alpha Actinin 4 | |
Western Blot 1 ug/ml, Immunocytochemistry/Immunofluorescence, Immunofluorescence 20 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
81 | |
IgG | |
Affinity purified | |
Antibody |
S06-3D7 | |
Western Blot, Immunofluorescence, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
A synthetic peptide of human Alpha Actinin 4 (Uniprot#: O43707) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title