Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Actinin 4 Recombinant Rabbit Monoclonal Antibody (5Y3R6)
GREENER_CHOICE

Catalog No. PIMA538061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538061 Supplier Invitrogen™ Supplier No. MA538061
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, 293T, Jurkat, Mouse brain, Mouse spleen, Rat lung Immunogen sequence: MVDYHAANQS YQYGPSSAGN GAGGGGSMGD YMAQEDDWDR DLLLDPAWEK QQRKTFTAWC NSHLRKAGTQ IENIDEDFRD GLKLMLLLEV ISGERLPKPE.

Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Actinin 4
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 5Y3R6
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Actn4
Gene Accession No. O43707, P57780, Q9QXQ0
Gene Alias actinin alpha 4; ACTININ-4; actinin-Alpha-4; Actn4; alpha actinin 4; alpha-actinin-4; C77391; F-actin cross-linking protein; focal segmental glomerulosclerosis 1; FSGS; FSGS1; non-muscle alpha-actinin 4
Gene Symbols Actn4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human alpha-Actinin-4 (O43707).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60595, 63836, 81
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.