Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Adaptin Antibody (AC1-M11), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300721MFV500
Description
alpha Adaptin Monoclonal antibody specifically detects alpha Adaptin in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
alpha Adaptin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Amphibian | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
AC1-M11 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin | |
100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
Purified bovine brain adaptor complexes. | |
0.1 mL | |
Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction | |
160 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction