Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha Adaptin Antibody (AP6), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001545AF647
Description
alpha Adaptin Monoclonal specifically detects alpha Adaptin in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| alpha Adaptin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 160 | |
| Human, Mouse, Rat, Bovine, Hamster, Primate | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| AP6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| AP2A1 | |
| Purified adaptors. | |
| 0.1 mL | |
| Primary | |
| Detects assembly polypeptide 2 (AP2) It recognizes the products of both alpha-adaptin genes, alpha A and alpha C as well as an alternatively spliced isoform of alpha A found in neurons. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction