Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Adaptin Monoclonal Antibody (AP6), Alexa Fluor™ 555, Invitrogen™

Catalog No. PIMA1064A55
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1064A55 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1064A55 Supplier Thermo Scientific Supplier No. MA1064A555

Mouse Monoclonal Antibody

alpha Adaptin Monoclonal antibody specifically detects alpha Adaptin in Bovine, Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Adaptin
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AP6
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 555
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH
Gene Ap2a1
Gene Accession No. O95782, P17426
Gene Alias 100 kDa coated vesicle protein A; Adaptor protein complex AP-2 subunit alpha-1; Adaptor-related protein complex 2 subunit alpha-1; Adtaa; ADTAB; Alpha1-adaptin; alpha2-adaptin; Alpha-adaptin A; alpha-adaptin C; AP2; AP-2 complex subunit alpha-1; Ap2a1; AP2-ALPHA; BOS_17797; Clapa1; CLAPA2; Clathrin assembly protein complex 2 alpha-A large chain; HIP9; HIP-9; HYPJ; Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit
Gene Symbols Ap2a1
Host Species Mouse
Immunogen Purified adaptors.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100772481, 11771, 160, 308578, 784636
Target Species Bovine, Hamster, Human, Mouse, Non-human Primate, Rat
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.