Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Adducin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577848 Supplier Invitrogen™ Supplier No. PA577848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Jurkat, Raji Predicted Reactivity: Rat (81%), Rhesus Monkey (100%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Adducins are a family of cytoskeleton proteins encoded by three genes (alpha, beta, gamma). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. Alpha- and beta-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. Alpha- and gamma-adducins are ubiquitously expressed. In contrast, beta-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Adducin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ADD1
Gene Accession No. P35611, Q9QYC0
Gene Alias Add1; ADD2; ADDA; ADDB; adducin 1; adducin 1 (alpha); Adducin beta; Adducin1; Adducin-1; Adducin-alpha; Adducin-beta; AI256389; Alpha adducin; alpha-ADD; alpha-adducin; beta Adducin; Beta-adducin; erythrocyte adducin alpha subunit; erythrocyte adducin subunit alpha
Gene Symbols ADD1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human alpha Adducin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11518, 118
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.