Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha',alpha'-Tetrafluoro-p-xylene 98.0+%, TCI America™
SDP

Catalog No. t17745g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 369-54-0
Molecular Formula C8H6F4
Molecular Weight (g/mol) 178.13
MDL Number MFCD01320708
InChI Key VWKMZVCSRVFUGW-UHFFFAOYSA-N
PubChem CID 2734031
IUPAC Name 1,4-bis(difluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)F)C(F)F

Specifications

Color Colorless
Boiling Point 70°C
Quantity 5 g
Synonym 1,4-bis difluoromethyl benzene, alpha,alpha,alpha',alpha'-tetrafluoro-p-xylene, acmc-1cr71, 1,4-bis-difluoromethyl-benzene, a,a,a',a'-tetrafluoro-p-xylene, benzene,1,4-bis difluoromethyl, benzene, 1,4-bis difluoromethyl, 1,4-bis bis fluoranyl methyl benzene, alpha,alpha,alpha,alpha-tetrafluoro-p-xylene
Formula Weight 178.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha,alpha,alpha′,alpha′-Tetrafluoro-p-xylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.