Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha,alpha-Trifluorotoluene, 99+%, ACROS Organics™

Click to view available options
Quantity:
50mL
500mL
2.5L
Packaging:
Plastic bottle
98-08-8
C7H5F3
146.11
MFCD00000372
GETTZEONDQJALK-UHFFFAOYSA-N
benzotrifluoride,trifluoromethyl benzene,alpha,alpha,alpha-trifluorotoluene,phenylfluoroform,benzene, trifluoromethyl,benzenyl fluoride,benzylidyne fluoride,usaf ma-16,a,a,a-trifluorotoluene,trifluoro toluene
7368
CHEBI:36810
trifluoromethylbenzene
C1=CC=C(C=C1)C(F)(F)F
98-08-8
C7H5F3
146.11
MFCD00000372
GETTZEONDQJALK-UHFFFAOYSA-N
benzotrifluoride,trifluoromethyl benzene,alpha,alpha,alpha-trifluorotoluene,phenylfluoroform,benzene, trifluoromethyl,benzenyl fluoride,benzylidyne fluoride,usaf ma-16,a,a,a-trifluorotoluene,trifluoro toluene
7368
CHEBI:36810
trifluoromethylbenzene
C1=CC=C(C=C1)C(F)(F)F
Infrared Spectrum Authentic
Refractive Index 1.4130 to 1.415
Melting Point -29°C
Boiling Point 102°C
Flash Point 12°C
Specific Gravity 1.19
Quantity 500mL
Color Colorless
Packaging Plastic bottle
Density 1.1900g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H5CF3
Beilstein 05, 290
Merck Index 15, 1113
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in many organic solvents
Formula Weight 146.11
Physical Form Liquid
Percent Purity ≥99.0%
Chemical Name or Material α, α, α-Trifluorotoluene, 99+%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-635-0

RTECSNumber : XT9450000

TSCA : TSCA