Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha,alpha'-Dibromo-m-xylene 97.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Melting Point | 76°C |
Color | White-Yellow |
Boiling Point | 140°C |
Quantity | 25 g |
UN Number | 2928 |
Formula Weight | 263.96 |
Percent Purity | ≥97.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | alpha,alpha′-Dibromo-m-xylene |
Safety and Handling
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction