Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,alpha-Dimethyl-gamma-butyrolactone 97.0+%, TCI America™
SDP

Catalog No. D273325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D273325G 25 g
D27335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D273325G Supplier TCI America Supplier No. D273325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3709-08-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD03844788
InChI Key UPVAIJPDWVTFKT-UHFFFAOYSA-N
Synonym Dihydro-3,3-dimethyl-2(3H)-furanone
PubChem CID 94746
IUPAC Name 3,3-dimethyloxolan-2-one
SMILES CC1(CCOC1=O)C

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 194°C
Quantity 25 g
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha,alpha-Dimethyl-gamma-butyrolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.