Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-Amanitin Monoclonal Antibody (3B9)
GREENER_CHOICE

Catalog No. PIMA549614
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549614 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549614 Supplier Invitrogen™ Supplier No. MA549614
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Alpha-amanitin is a cyclic peptide toxin found in certain species of mushrooms, including the death cap mushroom (Amanita phalloides). It is known for its potent inhibitory effects on RNA polymerase II, a key enzyme involved in transcription of genes. This inhibition can lead to liver failure and death in humans if ingested. Alpha-amanitin is also used in research as a tool to study gene expression and transcriptional regulation. It has been shown to be effective in killing cancer cells, and research is ongoing to explore its potential as a therapeutic agent for cancer treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Amanitin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 3B9
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of alpha-AMA hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.