Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Amylase 1 Monoclonal Antibody (c1), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543977
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543977 100 μL
PIMA543978 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543977 Supplier Invitrogen™ Supplier No. MA543977
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Amylase is an enzyme that catalyses the breakdown of starch into sugars. Amylase is present in human saliva, where it begins the chemical process of digestion. By in situ hybridization combined with high resolution cytogenetics, the amylase gene is mapped to 1p21. Amylase enzymes find use in bread making and to break down complex sugars such as starch (found in flour) into simple sugars. Yeast then feeds on these simple sugars and converts it into the waste products of alcohol and CO2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Amylase 1
Applications Western Blot
Classification Monoclonal
Clone c1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene AMY1A
Gene Accession No. P00687, P0DUB6
Gene Alias 1,4-alpha-D-glucan glucanohydrolase 1; alpha amylase 1; alpha-amylase 1; Amy1; Amy-1; Amy1a; Amy-1-a; AMY1B; AMY1C; AMY2A; amylase 1, salivary; amylase, alpha 1A (salivary); amylase, salivary, alpha-1A; C030014B17Rik; glycogenase; PA; salivary alpha-amylase; salivary amylase; salivary amylase alpha 1A; salivary and hepatic alpha-amylase
Gene Symbols Amy1, AMY1A
Host Species Mouse
Immunogen Ala15-Leu511 (Accession P0DUB6), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11722, 24203, 276
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.