Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Amylase 2B Monoclonal Antibody (2B12)
GREENER_CHOICE

Catalog No. PIMA518394
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518394 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518394 Supplier Invitrogen™ Supplier No. MA518394
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: PNTQQGRTSI VHLFEWRWVD IALECERYLA PKGFGGVQVS PPNENVAIHN PFRPWWERYQ PVSYKLCTRS GNEDEFRNMV TRCNNVGVRI YVDAVINHM.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Amylase 2B
Applications ELISA
Classification Monoclonal
Clone 2B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene AMY2B
Gene Accession No. P19961
Gene Alias 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase 2B; AMY2; Amy2-2; AMY2B; AMY3; amylase 2-2, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; OTTMUSG00000022462
Gene Symbols AMY2B
Host Species Mouse
Immunogen AMY2B (NP_066188, 19 a.a. ~ 117 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.