Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ alpha Amylase 2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112358
Description
alpha Amylase 2B Polyclonal Antibody for Western Blot, IHC (P), ELISA
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas.
Specifications
| alpha Amylase 2B | |
| Polyclonal | |
| Unconjugated | |
| AMY2B | |
| 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase 2B; AMY2; Amy2-2; AMY2B; AMY3; amylase 2-2, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; OTTMUSG00000022462 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 280, 545562 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19961 | |
| AMY2B | |
| Recombinant Human Alpha-amylase 2B protein (16-320AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction