Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha Amylase 2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100383
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100383 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100383 Supplier Invitrogen™ Supplier No. PA5100383
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AMY2B.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha Amylase 2B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AMY2B
Gene Accession No. P19961
Gene Alias 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase 2B; AMY2; Amy2-2; AMY2B; AMY3; amylase 2-2, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; OTTMUSG00000022462
Gene Symbols AMY2B
Host Species Rabbit
Immunogen A synthesized peptide derived from human AMY2B(Accession P19961), corresponding to amino acid residues A345-N395.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280, 545562
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.