Learn More
alpha Amylase Polyclonal Antibody, HRP, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $613.35
Specifications
Antigen | alpha Amylase |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | HRP |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1094
![]() ![]() |
Rockland Immunochemicals
20043450100 |
100 μg |
Each for $613.35
|
|
|||||
50-199-1095
![]() ![]() |
Rockland Immunochemicals
2004345S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Alpha Amylase Antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Rabbit Serum as well as purified and partially purified a-Amylase [Bacillus amyloliquefaciens]. Cross reactivity against a-Amylase from other tissues and species is unknown.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.Specifications
alpha Amylase | |
ELISA, Immunoprecipitation, Western Blot | |
HRP | |
Rabbit | |
Bacteria | |
1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; a-amylase; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a3; AMY2B; AMY3; amylase 2, pancreatic; amylase 2-2, pancreatic; amylase 2a1; amylase 2a3; amylase 2a3, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; Carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; mAmy2-4; mAmy2-6; OTTMUSG00000022430; OTTMUSG00000022432; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase; pancreatic amylase alpha 2A | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% gentamicin sulfate; pH 7.2 | |
Anti-Alpha Amylase Antibody was produced by repeated immunizations with Bacillus amyloliquefaciens a-Amylase protein. | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.