Learn More
Invitrogen™ alpha Amylase Polyclonal Antibody, HRP

Description
PA1-28946 detects alpha Amylase from bacteria samples.
Specifications
Specifications
| Antigen | alpha Amylase |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 10 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 |
| Gene Alias | 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; a-amylase; alpha-amylase; Alpha-amylase 2B; AMY2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a3; Amy2b; AMY3; amylase 2, pancreatic; amylase 2-2, pancreatic; amylase 2a1; amylase 2a3; amylase 2a3, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; mAmy2-4; mAmy2-6; OTTMUSG00000022430; OTTMUSG00000022432; OTTMUSG00000022462; PA; pancreatic alpha-amylase; pancreatic amylase; pancreatic amylase alpha 2A |
| Host Species | Rabbit |
| Immunogen | Bacillus amyloliquefaciens a-Amylase protein. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.