Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha Amylase Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990781
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0781 25 μL
50-199-0780 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0781 Supplier Rockland Immunochemicals Supplier No. 200401045S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Alpha Amylase Antibody is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified a-Amylase [Bacillus amyloliquefaciens]. Cross reactivity against a-Amylase from other tissues and species may occur but have not been specifically determined.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.

Specifications

Antigen alpha Amylase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Alias 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; a-amylase; alpha-amylase; Alpha-amylase 2B; Amy2; Amy2-1; Amy2-2; Amy2a; Amy2a1; Amy2a3; AMY2B; AMY3; amylase 2, pancreatic; amylase 2-2, pancreatic; amylase 2a1; amylase 2a3; amylase 2a3, pancreatic; amylase 2b; amylase 2b, pancreatic; amylase, alpha 2A (pancreatic); amylase, alpha 2B (pancreatic); Amy-X; Carcinoid alpha-amylase; glycogenase; HXA; mAmy2-1; mAmy2-4; mAmy2-6; OTTMUSG00000022430; OTTMUSG00000022432; OTTMUSG00000022462; PA; Pancreatic alpha-amylase; pancreatic amylase; pancreatic amylase alpha 2A
Host Species Rabbit
Immunogen a-Amylase [Bacillus amyloliquefaciens]
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.