Learn More
alpha-Angelicalactone, 98%
Description
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.
Solubility
Soluble in water (5g/100mL) and alcohol.
Notes
Moisture-sensitive. Store under inert gas. Protect from moisture. Incompatible materials are Oxidizing agents.
Specifications
Specifications
| Melting Point | 17°C to 18°C |
| Density | 1.093 |
| Boiling Point | 53°C to 54°C (10 mmHg) |
| Flash Point | 68°C (154°F) |
| Refractive Index | 1.448 |
| Quantity | 25 g |
| Beilstein | 108394 |
| Merck Index | 14,647 |
| Solubility Information | Soluble in water (5g/100mL) and alcohol. |
| Formula Weight | 98.1 |
| Show More |
Safety and Handling
| Hazard Category | H227 |
| Hazard Statement | GHS H Statement H227 Combustible liquid. |
| Precautionary Statement | P210-P280-P370+P378q-P501c |
| EINECSNumber | 209-701-8 |
| RTECSNumber | LU5075000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.