Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha B-crystallin Mouse anti-Human,Mouse,Bovine,Monkey,Rat, KHL, Clone: aB1788G7G6, MilliporeSigma™

Catalog No. MABN25522MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25522MI 25 μL
MABN25521MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25522MI Supplier MilliporeSigma Supplier No. MABN255225UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

alpha B-crystallin Monoclonal antibody specifically detects alpha B-crystallin in Human,Mouse,Bovine,Monkey,Rat samples. It is validated for ELISA, Immunofluorescence, Immunohistochemistry, Inhibition, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha B-crystallin
Applications ELISA, Immunofluorescence, Immunohistochemistry, Inhibition Assays, Western Blot
Classification Monoclonal
Clone aB1788G7G6
Conjugate KLH
Gene Accession No. NP_001276736
Gene Alias Alpha-crystallin B chain;Alpha(B)-crystallin;Heat shock protein beta-5;HspB5;Renal carcinoma antigen NY-REN-27;Rosenthal fiber component
Gene Symbols CRYAB, CRYA2, HSOB5
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal half of human Alpha(B)-crystallin.
Molecular Weight of Antigen ∼20 kDa observed; 20.16 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone aB1788G7G6 is a mouse monoclonal antibody that detects Alpha(B)-crystallin. It targets an epitope within 21 amino acids within the N-terminal half.
Target Species Human, Mouse, Bovine, Monkey, Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.